siRNA Details
| Gene Symbol: AW495222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045086.1 | 2 | 333 | |||
| GenBank mRNA | AK046172.1 | - | 2 | 334 | ||
| BB314344.2 | - | 2 | 156 | |||
| BB644732.1 | - | 2 | 331 | |||
| BE656108.1 | - | 2 | 216 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY712700.1 | - | 2 | 305 | Chr. 13 - 93706282 - 93719146 [-] on Build GRCm38 | |

AW495222
Genomic Map