siRNA Details
| Gene Symbol: Gm7056 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037571.1 | 2756 | ||||
| GenBank mRNA | AK135800.1 | - | 3 | 772 | ||
| AK136178.1 | - | 4 | 952 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm7056| Gene Symbol: Gm7056 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037571.1 | 2756 | ||||
| GenBank mRNA | AK135800.1 | - | 3 | 772 | ||
| AK136178.1 | - | 4 | 952 | |||