siRNA Details
| Gene Symbol: 4930562F07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038030.1 | 3 | 706 | |||
| NR_038031.1 | 3 | 634 | ||||
| GenBank mRNA | AK016191.1 | - | 3 | 707 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930562F07Rik| Gene Symbol: 4930562F07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038030.1 | 3 | 706 | |||
| NR_038031.1 | 3 | 634 | ||||
| GenBank mRNA | AK016191.1 | - | 3 | 707 | ||