siRNA Details
| Gene Symbol: 4930562M23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016193.1 | - | 1 | 393 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715800.1 | - | 1 | 390 | Chr. 13 - 109884222 - 109884866 [+] on Build GRCm38 | |

4930562M23Rik
Genomic Map