siRNA Details
| Gene Symbol: Gatsl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK033062.1 | - | 990 | |||
| AK035431.1 | - | 1006 | ||||
| BC026208.1 | - | 391 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gatsl2| Gene Symbol: Gatsl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK033062.1 | - | 990 | |||
| AK035431.1 | - | 1006 | ||||
| BC026208.1 | - | 391 | ||||