siRNA Details
| Gene Symbol: Gpr55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK032227.1 | - | 1 | 2115 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr55| Gene Symbol: Gpr55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK032227.1 | - | 1 | 2115 | ||