siRNA Details
| Gene Symbol: Gm15245 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK079668.1 | - | 4, 5 | 447 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB589581.2 | - | 4, 5 | 447 | Chr. X - 169447830 - 169475363 [+] on Build GRCm38 | |

Gm15245
Genomic Map