siRNA Details
| Gene Symbol: Ppp1r10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK004266.1 | - | 2 | 683 | ||
| AK011552.1 | - | 2 | 682 | |||
| AK031654.1 | - | 2 | 681 | |||
| AK079918.1 | - | 1 | 872 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp1r10| Gene Symbol: Ppp1r10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK004266.1 | - | 2 | 683 | ||
| AK011552.1 | - | 2 | 682 | |||
| AK031654.1 | - | 2 | 681 | |||
| AK079918.1 | - | 1 | 872 | |||