siRNA Details
| Gene Symbol: 2310030A07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040603.1 | 1, 2 | 103 | |||
| GenBank mRNA | AK009534.1 | - | 1, 2 | 104 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY709227.1 | - | 1, 2 | 104 | Chr. 1 - 150320681 - 150346771 [-] on Build GRCm38 | |

2310030A07Rik
Genomic Map