siRNA Details
| Gene Symbol: B830042I05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021034.1 | - | 1 | 105 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY727810.1 | - | 1 | 105 | Chr. 8 - 11075895 - 11076638 [+] on Build GRCm38 | |

B830042I05Rik
Genomic Map