siRNA Details
| Gene Symbol: C030002A05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021037.1 | - | 1 | 19 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY728097.1 | - | 1 | 19 | Chr. 8 - 17438796 - 17439150 [-] on Build GRCm38 | |

C030002A05Rik
Genomic Map