siRNA Details
| Gene Symbol: 2310065F04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038055.1 | 5 | 2655 | |||
| GenBank mRNA | AK010044.1 | - | 5 | 2655 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2310065F04Rik| Gene Symbol: 2310065F04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038055.1 | 5 | 2655 | |||
| GenBank mRNA | AK010044.1 | - | 5 | 2655 | ||