siRNA Details
| Gene Symbol: 1700057H21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006824.1 | - | 3 | 392 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706809.1 | - | 3 | 392 | Chr. 2 - 51421050 - 51437900 [-] on Build GRCm38 | |

1700057H21Rik
Genomic Map