siRNA Details
| Gene Symbol: Gssos1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK002817.1 | - | 2 | 666 | ||
| BY703321.1 | - | 2 | 665 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gssos1| Gene Symbol: Gssos1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK002817.1 | - | 2 | 666 | ||
| BY703321.1 | - | 2 | 665 | |||