siRNA Details
Gene Symbol: Tbc1d23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK028852.1 | - | 1 | 2540 | ||
AK045169.1 | - | 1 | 249 | |||
AK050825.1 | - | 1 | 386 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tbc1d23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK028852.1 | - | 1 | 2540 | ||
AK045169.1 | - | 1 | 249 | |||
AK050825.1 | - | 1 | 386 |