siRNA Details
| Gene Symbol: Tbc1d23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028852.1 | - | 1 | 2502 | ||
| AK045169.1 | - | 1 | 211 | |||
| AK050825.1 | - | 1 | 348 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbc1d23| Gene Symbol: Tbc1d23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028852.1 | - | 1 | 2502 | ||
| AK045169.1 | - | 1 | 211 | |||
| AK050825.1 | - | 1 | 348 | |||