siRNA Details
| Gene Symbol: Mid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY540027.1 | - | 476 | |||
| AY540028.1 | - | 476 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mid1| Gene Symbol: Mid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY540027.1 | - | 476 | |||
| AY540028.1 | - | 476 | ||||