siRNA Details
Gene Symbol: Gm33432 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK142623.1 | - | 54 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CJ065942.1 | - | 1 | 54 | Chr. 15 - 84127304 - 84135325 [+] on Build GRCm38 |
