siRNA Details
| Gene Symbol: Gm38554 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC049722.1 | - | 3 | 342 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU962724.1 | - | 3 | 345 | Chr. 5 - 119959243 - 119963370 [+] on Build GRCm38 | |

Gm38554
Genomic Map