siRNA Details
| Gene Symbol: 9330162012Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102323.1 | 2 | 892 | |||
| GenBank mRNA | AK034200.1 | - | 775 | |||
| AK080139.1 | - | 2 | 869 | |||
| BC094927.1 | - | 2 | 882 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9330162012Rik| Gene Symbol: 9330162012Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102323.1 | 2 | 892 | |||
| GenBank mRNA | AK034200.1 | - | 775 | |||
| AK080139.1 | - | 2 | 869 | |||
| BC094927.1 | - | 2 | 882 | |||