siRNA Details
| Gene Symbol: Gm35156 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK156776.1 | - | 3 | 2461 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm35156| Gene Symbol: Gm35156 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK156776.1 | - | 3 | 2461 | ||