siRNA Details
| Gene Symbol: 4833421G17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014745.1 | - | 1 | 57 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714242.1 | - | 1 | 57 | Chr. 1 - 87865073 - 87865893 [+] on Build GRCm38 | |

4833421G17Rik
Genomic Map