siRNA Details
| Gene Symbol: 4833422B07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014748.1 | - | 1 | 1284 | ||
| AV250295.1 | - | 1 | 113 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4833422B07Rik| Gene Symbol: 4833422B07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014748.1 | - | 1 | 1284 | ||
| AV250295.1 | - | 1 | 113 | |||