siRNA Details
| Gene Symbol: 3110037B15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014129.1 | - | 1 | 106 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY713114.1 | - | 1 | 106 | Chr. 9 - 112175087 - 112175465 [-] on Build GRCm38 | |

3110037B15Rik
Genomic Map