siRNA Details
Gene Symbol: 3110037B15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014129.1 | - | 1 | 227 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY713114.1 | - | 1 | 227 | Chr. 9 - 112175087 - 112175465 [-] on Build GRCm38 |
