siRNA Details
| Gene Symbol: A230005M16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK038413.1 | - | 1 | 1156 | ||
| AK046328.1 | - | 1 | 1151 | |||
| AK082064.1 | - | 2 | 1307 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A230005M16Rik| Gene Symbol: A230005M16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK038413.1 | - | 1 | 1156 | ||
| AK046328.1 | - | 1 | 1151 | |||
| AK082064.1 | - | 2 | 1307 | |||