siRNA Details
Gene Symbol: Slain1os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045148.1 | 1 | 37 | |||
GenBank mRNA | AK014964.1 | - | 1 | 48 | ||
AK029783.1 | - | 1 | 39 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slain1os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045148.1 | 1 | 37 | |||
GenBank mRNA | AK014964.1 | - | 1 | 48 | ||
AK029783.1 | - | 1 | 39 |