siRNA Details
| Gene Symbol: Slain1os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045148.1 | 3 | 1083 | |||
| GenBank mRNA | AK014964.1 | - | 3 | 1094 | ||
| AK029783.1 | - | 3 | 1085 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slain1os| Gene Symbol: Slain1os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045148.1 | 3 | 1083 | |||
| GenBank mRNA | AK014964.1 | - | 3 | 1094 | ||
| AK029783.1 | - | 3 | 1085 | |||