siRNA Details
| Gene Symbol: 2900057B20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045365.1 | 3 | 981 | |||
| GenBank mRNA | AK013712.1 | - | 3 | 984 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2900057B20Rik| Gene Symbol: 2900057B20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045365.1 | 3 | 981 | |||
| GenBank mRNA | AK013712.1 | - | 3 | 984 | ||