siRNA Details
| Gene Symbol: 2900057B20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045365.1 | 3 | 760 | |||
| GenBank mRNA | AK013712.1 | - | 3 | 763 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY712689.1 | - | 3 | 764 | Chr. 18 - 76091594 - 76098471 [-] on Build GRCm38 | |

2900057B20Rik
Genomic Map