siRNA Details
| Gene Symbol: 4930555M17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019767.1 | - | 2 | 348 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715739.1 | - | 2 | 348 | Chr. 3 - 5512993 - 5515333 [-] on Build GRCm38 | |

4930555M17Rik
Genomic Map