siRNA Details
Gene Symbol: 4930555B12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019765.1 | - | 2 | 230 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715730.1 | - | 2 | 230 | Chr. X - 114926989 - 114928602 [+] on Build GRCm38 |
