siRNA Details
| Gene Symbol: 4930555B12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019765.1 | - | 1 | 29 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715730.1 | - | 1 | 29 | Chr. X - 114926989 - 114928602 [+] on Build GRCm38 | |

4930555B12Rik
Genomic Map