siRNA Details
| Gene Symbol: 4930554H23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019764.1 | - | 1 | 838 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930554H23Rik| Gene Symbol: 4930554H23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019764.1 | - | 1 | 838 | ||