siRNA Details
| Gene Symbol: 4930552P12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045318.1 | 4 | 2108 | |||
| GenBank mRNA | AK019762.1 | - | 4 | 2109 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930552P12Rik| Gene Symbol: 4930552P12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045318.1 | 4 | 2108 | |||
| GenBank mRNA | AK019762.1 | - | 4 | 2109 | ||