siRNA Details
| Gene Symbol: 4930552P12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045318.1 | 2 | 295 | |||
| GenBank mRNA | AK019762.1 | - | 2 | 296 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715703.1 | - | 2 | 296 | Chr. 19 - 55642420 - 55702204 [-] on Build GRCm38 | |

4930552P12Rik
Genomic Map