siRNA Details
| Gene Symbol: 4930552P06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019761.1 | - | 1 | 396 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715702.1 | - | 1 | 396 | Chr. 4 - 107596780 - 107597226 [-] on Build GRCm38 | |

4930552P06Rik
Genomic Map