siRNA Details
| Gene Symbol: 4930549G23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045376.1 | 3, 4 | 340 | |||
| GenBank mRNA | AK016077.1 | - | 3, 4 | 342 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930549G23Rik| Gene Symbol: 4930549G23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045376.1 | 3, 4 | 340 | |||
| GenBank mRNA | AK016077.1 | - | 3, 4 | 342 | ||