siRNA Details
| Gene Symbol: 4930548K13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040656.1 | 3 | 524 | |||
| GenBank mRNA | AK016071.1 | - | 3 | 524 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715669.1 | - | 3 | 524 | Chr. 4 - 26635820 - 26705449 [-] on Build GRCm38 | |

4930548K13Rik
Genomic Map