siRNA Details
| Gene Symbol: 4930548K13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040656.1 | 2 | 259 | |||
| GenBank mRNA | AK016071.1 | - | 2 | 259 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930548K13Rik| Gene Symbol: 4930548K13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040656.1 | 2 | 259 | |||
| GenBank mRNA | AK016071.1 | - | 2 | 259 | ||