siRNA Details
| Gene Symbol: A030001D20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK003910.1 | - | 2 | 620 | ||
| AK037147.1 | - | 2 | 636 | |||
| AK049852.1 | - | 2 | 622 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A030001D20Rik| Gene Symbol: A030001D20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK003910.1 | - | 2 | 620 | ||
| AK037147.1 | - | 2 | 636 | |||
| AK049852.1 | - | 2 | 622 | |||