siRNA Details
| Gene Symbol: A030001D20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK003910.1 | - | 2 | 821 | ||
| AK037147.1 | - | 2 | 837 | |||
| AK049852.1 | - | 2 | 823 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY704165.1 | - | 2 | 821 | Chr. 7 - 45636217 - 45638742 [-] on Build GRCm38 | |

A030001D20Rik
Genomic Map