siRNA Details
| Gene Symbol: A530065N20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046142.1 | 3 | 867 | |||
| GenBank mRNA | AK080130.1 | - | 3 | 870 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A530065N20Rik| Gene Symbol: A530065N20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046142.1 | 3 | 867 | |||
| GenBank mRNA | AK080130.1 | - | 3 | 870 | ||