siRNA Details
| Gene Symbol: A530065N20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046142.1 | 2 | 193 | |||
| GenBank mRNA | AK080130.1 | - | 2 | 194 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB637326.1 | - | 2 | 194 | Chr. 13 - 60029711 - 60113578 [+] on Build GRCm38 | |

A530065N20Rik
Genomic Map