siRNA Details
| Gene Symbol: 1700010J16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040579.1 | 5 | 596 | |||
| GenBank mRNA | AK005834.1 | - | 5 | 596 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700010J16Rik| Gene Symbol: 1700010J16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040579.1 | 5 | 596 | |||
| GenBank mRNA | AK005834.1 | - | 5 | 596 | ||