siRNA Details
| Gene Symbol: 1700010J16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040579.1 | 2 | 113 | |||
| GenBank mRNA | AK005834.1 | - | 2 | 113 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU938461.1 | - | 154 | Chr. - - [-] on Build | ||

1700010J16Rik
Genomic Map