siRNA Details
| Gene Symbol: 1700010K23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040512.1 | 3 | 350 | |||
| GenBank mRNA | AK005838.1 | - | 3 | 352 | ||
| BY705986.1 | - | 3 | 352 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700010K23Rik| Gene Symbol: 1700010K23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040512.1 | 3 | 350 | |||
| GenBank mRNA | AK005838.1 | - | 3 | 352 | ||
| BY705986.1 | - | 3 | 352 | |||