siRNA Details
Gene Symbol: 1700010K23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040512.1 | 1 | 61 | |||
GenBank mRNA | AK005838.1 | - | 1 | 63 | ||
BY705986.1 | - | 1 | 63 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700010K23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040512.1 | 1 | 61 | |||
GenBank mRNA | AK005838.1 | - | 1 | 63 | ||
BY705986.1 | - | 1 | 63 |