siRNA Details
| Gene Symbol: Gm33045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK140017.1 | - | 3 | 435 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BF466703.1 | - | 2 | 186 | Chr. 13 - 110629173 - 110642633 [+] on Build GRCm38 | |

Gm33045
Genomic Map