siRNA Details
| Gene Symbol: Gm33045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK140017.1 | - | 5 | 919 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY655376.1 | - | 2 | 354 | Chr. 13 - 110638756 - 110642734 [+] on Build GRCm38 | |

Gm33045
Genomic Map